Drukuj
Lampy deuterowe
Dostawca: Agilent
2140-0590 G1314-60100 5182-1530
HEWL2140-0590EA 5620 PLN
HEWL2140-0590 HEWLG1314-60100 HEWL5182-1530
Lampy deuterowe
Oświetlenie Lampy deuterowe
These deuterium lamps are designed with a much narrower aperture providing increased light intensity and decreased noise, and tested for noise and drift specifications, correct operating voltage, light intensity and proper alignment.
  • Improved coating
  • Increasing lifetime to up to 2000 hours of use
  • Highly sensitive lamps, extends detection at trace levels
Certyfikaty: Manufactured in ISO 9001 environment and fully traceable, calibrated using optical standards provided by NIST, PTB.