Drukuj
Vacuum Controller, Vacuubrand
Dostawca: Vacuubrand
VACU20683201EA 0 PLN
VACU20683201 VACU20635983 VACU20635982
Vacuum Controller, Vacuubrand
Model CVC 3000
Fully electronic, highly versatile vacuum gauge and controller. The controller is equipped with a built-in capacitive absolute-pressure transducer made from alumina/ceramic as well as a built-in venting valve and switched outlets for a suction line valve and a coolant water valve. It eases work considerably and enables environmentally friendly operation whilst protecting samples, solvent recovery, reduced foam formation and superheating. The optimum vacuum setting can be found quickly and simply, even with unknown mixtures and pressures.
CVC 3000 + VSP 3000, KF DN 16 / CVC 3000 + VSP 3000, KF DN 25 / CVC 3000 + VACUU·VIEW extended, KF DN 16
The packages enable convenient vacuum control in the fine vacuum range down to 10-3 mbar and consist of a CVC 3000 vacuum controller, a VSP 3000 external Pirani vacuum sensor, a VV-B 15C in-line solenoid valve and all necessary small flange components. Small flanges KF DN 16, suitable for the VACUUBRAND RE/RZ 2.5 to RE/RZ 6 rotary vane pumps. Small flange KF DN 25 version suitable for our RE/RZ 9 to RE/RZ 16 rotary vane pumps. The CVC 3000 runs vacuum processes by control of vacuum, venting and cooling water valves. In these packages the vacuum control is performed by opening and closing the in-line solenoid valve VV-B 15C.
Dodatkowe informacje: Vacuum controller CVC 3000 (without internal vacuum sensor and venting valve) with VSP 3000, power supply, in-line solenoid valve VV-B 15C KF DN 16/ KF DN 25, T-piece KF DN 16/KF DN 25, hose nozzle DN 10 mm (PP) for KF DN 16, DN 15 mm (PP) for KF DN 25, clamping and sealing rings, ready for use, with operating manual.