Drukuj
Kolumny kapilarne, DB-624
Dostawca: Agilent
121-1324 122-1334 122-1364 123-1334 123-1364 123-1364E 124-1334 124-1374 125-1334 125-1334E 125-1364 125-1374 128-1324 122-1334E 122-1364E 123-1334E 125-1374E 125-1364E 128-1324E 121-1324E 121-1324LTM 122-1334LTM 123-1334LTM 124-1334LTM 125-13A4 125-13B4 125-1314 125-1334LTM 128-1314LTM 128-1324LTM 221-1324LTM 222-1334LTM
HEWL121-1324EA 4310 PLN
HEWL121-1324 HEWL122-1334 558-0364 558-0368 558-0369 HEWL123-1364E HEWL124-1334 HEWL124-1374 558-0372 HEWL125-1334E HEWL125-1364 HEWL125-1374 558-0375 HEWL122-1334E HEWL122-1364E HEWL123-1334E HEWL125-1374E HEWL125-1364E HEWL128-1324E HEWL121-1324E HEWL121-1324LTM HEWL122-1334LTM HEWL123-1334LTM HEWL124-1334LTM HEWL125-13A4 HEWL125-13B4 HEWL125-1314 HEWL125-1334LTM HEWL128-1314LTM HEWL128-1324LTM HEWL221-1324LTM HEWL222-1334LTM
Kolumny kapilarne, DB-624
Kolumny chromatograficzne
These columns are recommended for EPA methods 502.2 and 8021, as well as for fast GC/MS volatiles analysis. They are specifically designed for the analysis of volatile priority pollutants and residual solvents.
  • Excellent inertness for active compounds
  • Bonded and cross-linked
  • Solvent rinsable