Drukuj
Kolumny kapilarne, DB-35
Dostawca: Agilent
122-1932 122-1962 123-1932 123-1933 125-1912 125-1932 125-1937
HEWL122-1932EA 4940 PLN
HEWL122-1932 HEWL122-1962 HEWL123-1932 HEWL123-1933 HEWL125-1912 HEWL125-1932 HEWL125-1937
Kolumny kapilarne, DB-35
Kolumny chromatograficzne
They are solvent rinsable and very low bleed mid-polarity columns ideal for GC/MS. They are suitable for CLP pesticides.
  • Excellent inertness for active compounds
  • Enhanced mass spectral integrity
  • Bonded and cross-linked
Methylpolysiloxane
Max. temperature: 40 to 320/340 °C