Drukuj
Fiolki diagnostyczne, Nalgene®
Dostawca: Thermo Fisher Scientific
342035-0005 342035-0010 342035-0020
215-1166EA 7580 PLN
215-1166 215-1167 NALG342035-0020
Fiolki diagnostyczne, Nalgene®
Vialki
These clear PETG vials with white HDPE closures are ideal for sampling, storage and shipment of reagents or buffer solutions.
  • Non-pyrogenic and non-cytotoxic
  • Includes lined, white HDPE closure for a leakproof seal
Certyfikaty: Manufactured in compliance with ISO 13485:2003 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Sposób pakowania: Packed in shrink-wrapped trays with closures attached. These vials are bulk packed, 100 vials per tray, 500 vials per case.