Drukuj
Rotametry, Sigma-Aldrich
Dostawca: Merck
23320-U 23324 23326
SIAL23320-UEA 1420 PLN
SIAL23320-U SIAL23324 SIAL23326
Rotametry, Sigma-Aldrich
Przepływomierze
These rotameters are designed for accurate metering and chromatographic purposes.
  • Flow tube and four interchangeable floats
  • Standard flow tables (ml/minute at STP) included
  • Can be mounted or installed on the (optional) tripod assembly
Suitable for chromatography applications.