Drukuj
Kolby miarowe, klasa A, Corning®
Dostawca: Corning
5640P-500 5640P-1L 5640P-50 5640P-100 5640P-25 5640P-10 5640P-250
CORN5640P-500EA 1080 PLN
CORN5640P-500 CORN5640P-1L CORN5640P-50 CORN5640P-100 CORN5640P-25 CORN5640P-10 CORN5640P-250
Kolby miarowe, klasa A, Corning®
Kolby Kolby miarowe
PMP, przezroczyste, ze stożkowym korkiem PP.
  • Mogą być autoklawowane w 121°C bez wpływu na błedy wskazań
  • Rutynowe czyszczenie w temperaturze do 60°C nie wpływa na błędy wskazań
  • With an individually calibrated graduation line
The accuracy of volumetric ware depends on the care used in calibrating it, using the correct type of ware for the application, handling the ware properly and ensuring the ware is clean. Calibration of volumetric ware is usually done at 20 °C, and the ware should be used at approximately this temperature. Refrigerated liquids should be allowed to come to room temperature before measuring them. Under normal use and care, the calibration of volumetric ware will not change. Do not expose volumetric ware to excessive heat, defined as temperatures approaching the Corning-defined upper service limit. The polymers from which the Corning line is manufactured have unique thermal characteristics. The polymer temperature capability is described above.
It is important to recognise the difference in specifications regarding DIN and ASTM standards. For reference, ASTM Class A and B specifications are more precise than DIN Class A and B specifications, respectively.
Certyfikaty: Klasa A w nawiązaniu do DIN EN ISO 1042.
Dodatkowe informacje: Produkty będą dostarczane z nadrukowanym numerem partii i certyfikatem.