Drukuj
Opaski kablowe, nierozrywalne, Allplastik-Kabelbinder®
Dostawca: WUERTEMBERGISCHE ALLPLASTIK
5203 5211 5216 5223 5226 5217
129-4814EA 853 PLN
129-4814 129-4815 129-4816 129-4817 129-4818 WUER5217
Opaski kablowe, nierozrywalne, Allplastik-Kabelbinder®
Pasy
These Allplastik-Kabelbinder® cable ties are made of high-quality polyamide 6.6, have high tensile strength and are available in different lengths.
  • With a curved round tip to make it easier to thread into the eye of the cable tie
  • Rounded edges for safe usage
  • Material with minimal coefficients of friction
  • Printing or laser engraving is possible
  • Temperature resistance: −40 to +85 °C
The ties are ideal for electrical installations, for wiring in industrial systems and control boards, for the automobile sector.
Certyfikaty: IEC EN 62275.