Drukuj
Filtry z wkładem, ProcessGard® CN
Dostawca: ENTEGRIS
CN0601E06 CN1H01E06 CN5001E06 CN5002E06
ENTECN0601E06EA 0 PLN
ENTECN0601E06 ENTECN1H01E06 ENTECN5001E06 ENTECN5002E06
Filtry z wkładem, ProcessGard® CN
Filtry Wkłady filtracyjne
These cartridge filters are designed for the removal of particles and microorganisms from liquids and gases. Used in clarification and prefiltration applications, these pleated filters retain contaminants primarily on the surface of the filter media. Downstream filter performance is enhanced due to efficient particle and microorganism reduction.
  • Removal of silica particles, carbon fines, resin fines and other particulates and microorganisms
  • Excellent chemical and heat resistance
  • Manufactured, tested and packaged in an ISO 9001 Quality Systems Standard facility
Membrane: Non woven PP; Supports, cage, core, sleeve and endcaps: PP
Maximum differential pressure: 4,83 bar at 25 °C
Maximum operating temperature: 80 °C