Drukuj
Nośniki płytek, do obsługi płytek 100 mm (4"), serie 72/182 i 190
Dostawca: ENTEGRIS
A72-40MB-0215 A18239MLB-0215 PA7240MB-61C02 A182-39M-0215 PA18239MLB61C2 A190-40M-0215 PA18239M-61C02 PA72-40MB-0603 PA182-39M-0603 A182-39MU-0215 A7239M060215 PA18239MLC0603 PA18239MLC61C0
216-1632EA 0 PLN
216-1632 ENTEA18239MLB-0215 216-1697 216-1571 216-1674 216-1574 ENTEPA18239M-61C02 ENTEPA72-40MB-0603 ENTEPA182-39M-0603 ENTEA182-39MU-0215 216-1631 ENTEPA18239MLC0603 ENTEPA18239MLC61C0
Nośniki płytek, do obsługi płytek 100 mm (4"), serie 72/182 i 190
Urządzenia do wafli Transportery do płytek krzemowych
These 100 mm wafer carriers are designed for optimum performance in automated operations or for processing LCDs and other small devices.
72 series
Original linear wafer carrier
Wash openings
'H' bar equipment interface
Pins and holes on top rail
Both end walls have flanges that extend to the bottom of carrier
182 series
Open sidewall style with wrap-around wash slots
Protective side wall design guards against damage from operator contact
190 series
Optimum performance in centrifugal processing applications
Low contour, open flow sidewall style
'H' bar equipment interface
Pins and holes on top rail
Uwaga: Articles may not be available in all countries. Please contact your local Avantor office.