Drukuj
Nośniki płytek do transportu płytek 150 mm, seria 182/194/196
Dostawca: ENTEGRIS
PA18260MB61C02 PA18260MB-0603 PA19460MB-0603 PA19460MB61C02 PA18260M-61C02 PA18260MC61C02 K19660MLB97C02
216-1693EA 0 PLN
216-1693 216-1692 216-1695 216-1696 216-1691 ENTEPA18260MC61C02 ENTEK19660MLB97C02
Nośniki płytek do transportu płytek 150 mm, seria 182/194/196
Urządzenia do wafli Transportery do płytek krzemowych
These standard 150 mm wafer carriers feature a high rigidity and dimensional stability.
  • Open sidewall design to minimize liquid traps, to promote even solution coverage and drainage
  • High profile design for maximum wafer protection
  • Capacity of 25 wafers
182 series - Industry standard wafer carrier
Four-point contact on the 'H' bar for improved wafer positioning (only some articles)
Open sidewall style
High profile design for maximum wafer protection
Pins and holes on the top rail
194/196 series - Superior automation performance
Four-point contact on the 'H' bar for improved wafer positioning
Open sidewall style
Locating track notches at the centre of the 13th pocket
'H' bar equipment interface
Pins and holes on the top rail
Uwaga: Articles may not be available in all countries. Please contact your local Avantor office.