Drukuj
Środki dezynfekujące, ClearKlens Tego® 2000
Dostawca: Diversey
7516051 7514341 7513387 7516427
141-0891EA 1760 PLN
141-0891 DIVY7514341 141-1220 141-1227
Środki dezynfekujące, ClearKlens Tego® 2000
Środki czystości Środki do dezynfekcji powierzchni
Highly effective, active amphoteric, surface disinfectant with a broad spectrum of antimicrobial activity, developed for use in sterile environments.
  • Ready to use, 0,2 µm filtered, sterile filled and double-bagged
  • Validated HPLC and titration test methods available
  • Compatible with ClearKlens Cleansinald and ClearKlens Bi-Spore as a rotational process
Certyfikaty: Passes CEN 1276, CEN 1650 and CEN 13697
Uwaga: Use biocides safely. Always read the label and product information before use.
Articles may not be available in all countries. Please contact your local Avantor office.