Drukuj
Obuwie zawodowe ESD, sandały, Nature, 4085
Dostawca: Abeba
4085/34 4085/35 4085/36 4085/37 4085/38 4085/39 4085/40 4085/41 4085/42 4085/43 4085/44 4085/45 4085/46 4085/47 4085/48
ABEB4085/34EA 295 PLN
ABEB4085/34 ABEB4085/35 ABEB4085/36 ABEB4085/37 ABEB4085/38 ABEB4085/39 ABEB4085/40 ABEB4085/41 ABEB4085/42 ABEB4085/43 ABEB4085/44 ABEB4085/45 ABEB4085/46 ABEB4085/47 ABEB4085/48
Obuwie zawodowe ESD, sandały, Nature, 4085
Obuwie i buty
ESD sandals made from smooth leather and a rubber sole (resistant to fuel, oil, acids and bases).
  • Skin friendly leather coated, cork integrated insole with toe gripper
  • Sanitized® treatment reduces odour and bacterial growth
  • Slip resistant special rubber sole with shock-absorbent heel
  • Adjustable instep area with Velcro® fastening
Colour: Black
Certyfikaty: EN 61340 (ESD); TÜV GS DIN 4843/100 § 3.1v.8/93, EN ISO 20347:2012; OB, FO, A, SRC