Drukuj
VWR® Q Path® Microtwin, kasety, w dozowniku
Dostawca: VWR Collection
720-2400EA 1400 PLN
720-2400 720-2401 720-2402 720-2403 720-2404 720-2405 720-2406 720-2407 720-2408
VWR® Q Path® Microtwin, kasety, w dozowniku
Kasety do osadzania Kasety biopsyjne
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassette with separated disposable lid, supplied in racks with 40 cassettes for Leica and Sakura cassette printers.
  • No biopsy pads, biopsy capsules or filter paper needed
  • Surface area of pore: 0,3×0,3 mm²
  • Hinge
  • 45° writing angle