Drukuj
Maski pełnotwarzowe, 3S
Dostawca: M S A
2055718 D2055000 2055-779
111-1915EA 0 PLN
111-1915 111-1565 111-2333
Maski pełnotwarzowe, 3S
Maski oddechowe Maski wielokrotnego użytku Maski pełnotwarzowe
These reusable full face masks with a soft mask body made from a special rubber compound or silicone and a lens made from impact resistant PC offers good visibility and low breathing resistance.
  • The wide sealing edge and a deep chin cup ensure a snug, comfortable fit and a tight seal
  • Large field of vision for excellent visibility
  • Speech membrane in the connector ensures good communication
  • Exhalation valve disc and valve housing are easily replaceable
  • Head strap enables the mask to be put on and removed quickly and ensures a pressure-free, secure fit
  • Inner mask ensures optimum airflow and prevents the lens from fogging
The mask has a standard thread connection and has been developed for use with filters, normal pressure compressed air breathing apparatus, airline breathing devices and power-assisted filtering devices.
Certyfikaty: EN 148-1, EN 136 (Class 3).
Uwaga: *One size fits all