Drukuj
Refraktometry ręczne, seria HR
Dostawca: A.KRÜSS Optronic
HR181 HRT32 HRN62 HRN82 HR92 HRO32 HRM18 HRMT18 HR27/100 HRS16 HR146 HR25/800 HR900 HR901 HRKFZ1 HROT32 HRKL32 HRB32-T HRB92-T
635-2601EA 0 PLN
635-2601 635-2603 635-2604 635-2606 635-2607 635-2608 635-2609 635-2610 635-2611 635-2612 635-2613 635-2614 635-2615 635-2616 635-2618 KRUEHROT32 KRUEHRKL32 KRUEHRB32-T KRUEHRB92-T
Refraktometry ręczne, seria HR
Refraktometry Refraktometry przenośne
HR series models possess various scales and additional functions which ensures the appropriate handheld refractometer for many application areas.
  • Reliability when reading, as the measured value does not first have to be converted
  • Selected models have an automatic temperature compensation which increases measurement precision for measurements which are performed at 10 to 40 °C instead of 20 °C
  • For calibration, distilled water is required, or else a small calibration plate is provided
These refractometers are useful for the determination of mixing ratios, quality and quantity inspection in the beverages, food, sugar, chemicals, flavours, petrochemicals, cosmetics, metalworking, pharmaceuticals, water, education and research domains.