Drukuj
Bandaże, Aluderm
Dostawca: SOEHNGEN
1003372 1003373 1003371
118-1243EA 51.3 PLN
118-1243 118-1244 SOEH1003371
Bandaże, Aluderm
Artykuły pierwszej pomocy Opatrunki i plastry
Aluminised wound dressing made of polyester non woven fabric. The final layer is a hydrophobic PP non woven fabric. These bandages feature a no-touch winding.
  • Absorbent layer has an absorption capacity of 1400 g/m²
  • Will not stick to any wound, breathable
  • Bandages do not release fibres
  • Strongly absorbent, keeping even the highly secreting wounds dry
  • Promotes skin formation and healing, physiologically safe, free of chemical binding materials