Drukuj
Złącza węży, Nalgene
Dostawca: Thermo Fisher Scientific
7210-0500 7211-0500
NALG7210-0500EA 1160 PLN
NALG7210-0500 NALG7211-0500
Złącza węży, Nalgene
Okucia i złącza
PP, white.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable
Mini hose barb sanitary adapter fitting for carboys and tanks.
Certyfikaty: Biologically compatible when tested according to the U.S. Pharmacopoeia XXII