Drukuj
Zbiorniki bębnowe, PE-200-EP
Dostawca: LACONT UMWELTTECHNIK
P70-7010-A P70-7012-A P70-7014-A P70-7025-A P70-7027-A P70-7011-A P70-7015-A P70-7023-A P70-7024-A P70-7026-A P70-7061-A P70-7062-A P70-7058-A P70-7059-A P70-7030-A P70-7020-A P70-7021-A P70-7022-A P70-7013-A P70-7029-A P70-7028-A
LACOP70-7010-AEA 0 PLN
LACOP70-7010-A LACOP70-7012-A LACOP70-7014-A LACOP70-7025-A LACOP70-7027-A LACOP70-7011-A LACOP70-7015-A LACOP70-7023-A LACOP70-7024-A LACOP70-7026-A LACOP70-7061-A LACOP70-7062-A LACOP70-7058-A LACOP70-7059-A LACOP70-7030-A LACOP70-7020-A LACOP70-7021-A LACOP70-7022-A LACOP70-7013-A LACOP70-7029-A LACOP70-7028-A
Zbiorniki bębnowe, PE-200-EP
Składowanie beczek
Ideal for storing water-endangering and aggressive media.
  • Rotationally-molded body of black PE
  • Circumferential stabilising ribs
  • Ground clearance for internal transport
  • Galvanised or PE grid support