Drukuj
Pojemniki transportowe bezpieczeństwa
Dostawca: LACONT UMWELTTECHNIK
L15-5010-A L15-5011-A L15-5012-A L15-5013-A L15-5014-A
LACOL15-5010-AEA 0 PLN
LACOL15-5010-A LACOL15-5011-A LACOL15-5012-A LACOL15-5013-A LACOL15-5014-A
Pojemniki transportowe bezpieczeństwa
Pojemniki transportowe Nosidła
Container and push-over lid both made of PE, with metal clamps for closing.
Suitable for the transport of hazardous good such as paints and spray cans.
Certyfikaty: Complies with ADR, RID, IMDG.