Drukuj
Szafki do przechowywania butli z gazem pod ciśnieniem, TRG
Dostawca: LACONT UMWELTTECHNIK
C27-3300-B C27-3301-B C27-3302-B
LACOC27-3300-BEA 0 PLN
LACOC27-3300-B LACOC27-3301-B LACOC27-3302-B
Szafki do przechowywania butli z gazem pod ciśnieniem, TRG
Meble Szafy bezpieczeństwa
Storage cabinets are used for the storage of pressurised gas cylinders indoors and outdoors.
  • High corrosion protection
  • Stainless steel floor to prevent impact damage
  • Ventilation by means of ventilation slots in the doors and below the roof
  • Doors with cylinder lock and roof inclined towards front
  • DN 80 cabinet roof prepared for on-site technical forced ventilation system
  • Lockable
Outer body made from galvanised sheet steel, powder-coated in RAL 7035 - light grey. Doors completely made from sheet steel or sheet steel door with plexi glass cut-out.
It can also be used indoors depending on defined fire load. Storage and operation even without compliance of protection zones.
Certyfikaty: Conforms with TRG 700, TRG 1050, TRG 1400, TRGs 510, ASR A1,3.
Dodatkowe informacje: Fully equipped TRG cells include 2× vertical c-rails on back wall, 2× height- adjustable, horizontal c-rails for mounting fittings, 1× cylinder holder (tension belt) for 50 L gas cylinders, outlets in roof for pipe penetration (Ø 25 mm) including filler plug.