Drukuj
Obuwie ochronne ESD, chodaki, HACCP, Anatom, 32120/32125
Dostawca: Abeba
32120/39 32120/41 32120/43 32120/45 32120/46 32120/38 32120/40 32120/44 32120/36 32120/37 32120/42 32120/47 32120/48 32125/36 32125/37 32125/38 32125/39 32125/40 32125/41 32125/42 32125/43 32125/44 32125/45 32125/46 32125/47 32125/48 32120/49 32120/50 32120/51 32125/49 32125/50 32120/52 32125/51
ABEB32120/39EA 405 PLN
ABEB32120/39 ABEB32120/41 ABEB32120/43 ABEB32120/45 ABEB32120/46 ABEB32120/38 ABEB32120/40 ABEB32120/44 ABEB32120/36 ABEB32120/37 ABEB32120/42 ABEB32120/47 ABEB32120/48 ABEB32125/36 ABEB32125/37 ABEB32125/38 ABEB32125/39 ABEB32125/40 ABEB32125/41 ABEB32125/42 ABEB32125/43 ABEB32125/44 ABEB32125/45 ABEB32125/46 ABEB32125/47 ABEB32125/48 ABEB32120/49 ABEB32120/50 ABEB32120/51 ABEB32125/49 ABEB32125/50 ABEB32120/52 ABEB32125/51
Obuwie ochronne ESD, chodaki, HACCP, Anatom, 32120/32125
Obuwie i buty
ESD shoe made from microfibre with a non foldable, adjustable heel strap and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Perspiration-absorbent special lining with silver fibre
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Asymmetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock-absorbent PU midsole
  • Protective steel toecap (200 J)
  • Washable at 30 °C
Suitable for hygienically demanding areas.
Colour: White/black.
Certyfikaty: EN 61340 (ESD); DGUV 112-191; HACCP; EN ISO 20345:2011; SB.