Drukuj
Akcesoria do statywów, RSG solingen
Dostawca: Kango
7KA80XHFO00512 7KA80YHFO00905 7NG35GDLR00905 7NG35GDLR02905 7NG35GDLU02905 7NG35GDLV00905 7NG40GBLU00512 7NG20GHLP02905 7NG40GBLR00512 7NG35NBPP00905 7NG35GHLP02905 7NG35GHLR00905 7NG35GHLR02512 7NG35GHLR02905 7NG35GHLU02905 7NG35GTLP01905 7NG35GDLV02512 7NG35NBPR00512 7NG35NBPR00905 7NG35NHPP00905 7NG35NHPP02905 7NG35NHPR02905 7NG40GBLP00905 7NG35GHLV02905 7NG35GHLP00905 7NG35GDLV02905 7NG20GHLP00512 7NG20GHLP02512 7NG20NHNP02905 7NG35GBLC00512 7NG35GBLC00905 7NG35GBLP00905 7NG35GBLR00512 7NG35GBLR00905 7NG20GHLP00905 7NG35GBLU00905 7NG40GBLU00905 7NG40GDLP01905 7NG40GHLP00905 7NG40GHLP01512 7NG40GHLP01905 7NG40GBLR00905 7NG20NHNP00905 7NG35GHLU02512 7NG35GTLP01512
139-0438EA 1610 PLN
139-0438 139-1010 139-0720 139-0721 139-0722 139-0723 139-0739 BIEN7NG20GHLP02905 139-0738 139-0733 139-0697 139-0726 139-0727 139-0728 139-0730 139-0732 139-0724 BIEN7NG35NBPR00512 139-0734 139-1014 139-0735 139-0736 139-0737 139-0731 139-0725 139-1013 BIEN7NG20GHLP00512 BIEN7NG20GHLP02512 BIEN7NG20NHNP02905 139-0715 139-0716 139-0717 139-0718 139-1012 BIEN7NG20GHLP00905 139-0719 139-0740 139-0741 139-0742 139-0743 139-1016 139-1015 BIEN7NG20NHNP00905 139-0729 BIEN7NG35GTLP01512
Akcesoria do statywów, RSG solingen
Krzesła
Ergonomic seats in soft polyurethane for maximum comfort. Soft PU, suitable for many uses and work environments.
  • Does not overheat or cause perspiration
  • Very sturdy: This range resists sharp tools, sparks, weak acids and hydrocarbons
  • Easily cleaned using common household products
  • Foam thickness: 4 cm
Chairs:
Black or blue ergonomic square seat and curved backrest for better lumbar support
Incorporated carry handle
Articulated backrest adjustable in height and depth (standard chairs)
Independently adjustable seat and backrest tilt with one fingertip lever thanks to an asynchronous mechanism (asynchronous chairs)
Dimensions of the seat (W×D): 410×390 mm
Dimensions of the backrest (W×H): 360×220 mm
Gas lift column with shock absorber for height adjustment
5-branch scuff resistant reinforced base
Semi stand-up seats:
Black or blue ergonomic square seat
Dimensions of the seat (W×D): 410×390 mm
Gas lift column with schock absorber for height adjustment
5-branch scuff resistant reinforced base
Stand-up seats:
Ergonomic concept that eliminates the fatigue created by body weight while standing up
Extremely stable and easy to move
Space saving
Its base is designed not to interfere with user movements
Semicircular steel base covered in PVC, plastic based satin coating, floor space (W×D): 490×330 mm
Dimension of the seat (W×D): 345×175 mm
Non rotating, tilted gas lift column with height adjustment and shock absorber
* 750 to 980 mm with extension piece.
Informacje do zamówienia: The whole seat range are classified CF according to the recommendation D2 2000 of GPEM, April 2000.