Drukuj
Trousers for women, C.I., Design C, royal blue
Dostawca: FRISTADS KANSAS
844530131C44 844530131C46 844530131C48 844530210C44 844530210C46 844530210C48 844530231C44 844530231C46 844530231C48 844530331C44 844530331C46 844530331C48 844530440C44 844530440C46 844530440C48 844530520C44 844530520C46 844530520C48 844530530C44 844530530C46 844530530C48 844530539C44 844530539C46 844530539C48 844530640C44 844530640C46 844530640C48 844530730C44 844530730C46 844530730C48 844530940C44 844530940C46 844530940C48 844530941C44 844530941C46 844530941C48 84453034 84453036 84453038 84453040 84453042 84453044 84453046 84453048 84453050
KWIN844530131C44EA 0 PLN
KWIN844530131C44 KWIN844530131C46 KWIN844530131C48 KWIN844530210C44 KWIN844530210C46 KWIN844530210C48 KWIN844530231C44 KWIN844530231C46 KWIN844530231C48 KWIN844530331C44 KWIN844530331C46 KWIN844530331C48 KWIN844530440C44 KWIN844530440C46 KWIN844530440C48 KWIN844530520C44 KWIN844530520C46 KWIN844530520C48 KWIN844530530C44 KWIN844530530C46 KWIN844530530C48 KWIN844530539C44 KWIN844530539C46 KWIN844530539C48 KWIN844530640C44 KWIN844530640C46 KWIN844530640C48 KWIN844530730C44 KWIN844530730C46 KWIN844530730C48 KWIN844530940C44 KWIN844530940C46 KWIN844530940C48 KWIN844530941C44 KWIN844530941C46 KWIN844530941C48 KWIN84453034 KWIN84453036 KWIN84453038 KWIN84453040 KWIN84453042 KWIN84453044 KWIN84453046 KWIN84453048 KWIN84453050
Trousers for women, C.I., Design C, royal blue
Odzież robocza
Robust trousers for ladies, made from 65% polyester and 35% cotton, with soft nap on reverse side.
  • Fly with zip
  • Two front pockets, two back pockets with flap and velcro closure
  • Leg pockets with flaps and three tool pockets, leg pocket with mobile phone pocket and two pen pockets
  • Folding-ruler pocket, D-ring, inside knee pockets
Weight: 260 g/m²
Colour: Royal blue
Informacje do zamówienia: Trousers are available without any coloured accents or with 12 different accent colours.