Drukuj
Płytki PCR, 96-dołkowe, bez osłony, niski profil, Axygen®
Dostawca: Corning
PCR-96-LP-FLTW PCR-96LP-FLT-C
732-1022EA 570 PLN
732-1022 732-1024
Płytki PCR, 96-dołkowe, bez osłony, niski profil, Axygen®
Mikropłytki Płytki do PCR
PP plates, silicone mats. Designed to fit 0,2 ml thermal cycler blocks.
  • Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified RNase, DNase and endotoxin-safe for highest sample integrity
  • AxyMat™ sealing mats autoclavable and reusable
Volume with AxyMat™: 106 μl
Compatible with (in addition to standard thermal cycling): Eppendorf Realplex, MJ Opticon, Chromo4 qPCR, Stratagene QPCR.