Drukuj
Systemy regałów na ciecze zagrażające wodzie i agresywne, KG-GR WPE
Dostawca: LACONT UMWELTTECHNIK
R71-2120-D R71-2121-D R71-2122-D R71-2123-D
LACOR71-2120-DEA 0 PLN
LACOR71-2120-D LACOR71-2121-D LACOR71-2122-D LACOR71-2123-D
Systemy regałów na ciecze zagrażające wodzie i agresywne, KG-GR WPE
Meble Regały i szafki
Shelving systems are used for the storage of water-endangering and aggressive liquids.
  • Galvanised shelving parts and tray-shaped shelves with PE insert
  • Height-adjustable shelves at intervals of 25 mm
  • Mixture of basic and extension units allows extension as required
  • Stable construction by reinforced struts
Two different combinable shelf widths (1000 and 1300 mm) allow optimal utilisation of existing storage space.
Maximum unit load: 1200 kg.