Drukuj
Systemy regałów na ciecze zagrażające wodzie i agresywne, KG-GR
Dostawca: LACONT UMWELTTECHNIK
R71-2130-D R71-2133-D R71-2140-D R71-2143-D R71-2131-D R71-2132-D R71-2141-D R71-2142-D
LACOR71-2130-DEA 0 PLN
LACOR71-2130-D LACOR71-2133-D LACOR71-2140-D LACOR71-2143-D LACOR71-2131-D LACOR71-2132-D LACOR71-2141-D LACOR71-2142-D
Systemy regałów na ciecze zagrażające wodzie i agresywne, KG-GR
Meble Regały i szafki
Shelving systems are used for the storage of water-endangering and aggressive liquids.
  • Galvanised shelving parts and tray-shaped shelves with PE insert
  • Height-adjustable shelves at intervals of 25 mm
  • Mixture of basic and extension units allows extension as required
  • Stable construction by reinforced struts
Two different combinable shelf widths (1000 and 1300 mm) allow optimal utilisation of existing storage space.
Maximum unit load: 1200 kg.