Drukuj
Palety i wanny do bębnów, CW
Dostawca: LACONT UMWELTTECHNIK
P65-2001-A P65-2002-A P65-2003-A P65-2004-A P65-2005-A P65-1001-A P65-1002-A P65-1003-A P65-1004-A P65-1005-A
LACOP65-2001-AEA 0 PLN
LACOP65-2001-A LACOP65-2002-A LACOP65-2003-A LACOP65-2004-A LACOP65-2005-A LACOP65-1001-A LACOP65-1002-A LACOP65-1003-A LACOP65-1004-A LACOP65-1005-A
Palety i wanny do bębnów, CW
Składowanie beczek
These pallets and sumps made from 3 mm sheet steel are ideal for the storage of water-endangering and flammable liquids.
  • Removable galvanised grid
  • Ground clearance 100 mm for internal transport
  • Corrosion protection by primer coating and painting or by fire-galvanised sheet steel