Drukuj
Palety i wanny do KTC, WSP
Dostawca: LACONT UMWELTTECHNIK
P21-2602-B P21-2607-A P21-1602-B P21-1607-A P21-2601-B P21-1601-B P21-2605-A P21-1605-A
LACOP21-2602-BEA 0 PLN
LACOP21-2602-B LACOP21-2607-A LACOP21-1602-B LACOP21-1607-A LACOP21-2601-B LACOP21-1601-B LACOP21-2605-A LACOP21-1605-A
Palety i wanny do KTC, WSP
Składowanie beczek
These pallets and sumps made of 3 mm sheet steel are ideal for storing and filling off hazardous substances from large containers.
  • Sump capacity of 1000 L
  • Support surface is a removable fire-galvanised grid
  • Ground clearance 100 mm for fork-lift truck transport
  • High corrosion resistance – with primer and painting or fire galvanisation