Drukuj

Bufory, glicyloglicyna, 0,2 M roztwory, Thermo Fisher Scientific

Dostawca: Thermo Fisher Scientific

J60717.AE J60717.AK J62920.AK
J60717.AEEA 374 PLN
J60717.AE J60717.AK J62920.AK
Bufory, glicyloglicyna, 0,2 M roztwory, Thermo Fisher Scientific
Bufory Bufory biologiczne

CAS No.: 556-50-3
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00008130

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR