2-Formylimidazole ≥97%
Dostawca: Thermo Fisher Scientific
|
Uwaga
|
Synonim:
1H-Imidazole-2-carbaldehyde, Imidazole-2-carboxaldehyde
L13883.06
L13883.14
L13883.06EA
824
PLN
L13883.06
L13883.14
2-Formylimidazole ≥97%
2-Formylimidazole
Purity: 97%
CAS No.: 10111-08-7
Molecular Formula: C4H4N2O
Formula Weight: 96.09
Storage Temperature: 2°C to 8°C
Melting Point: ca 205° dec.
Beilstein Reference: 4371302
MDL No.: MFCD00003544
Formuła:
C₄H₄N₂O |
Numer MDL:
MFCD00003544 Numer CAS: 10111-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...