Drukuj
Kombinezon do pomieszczeń czystych z mankietami na nogawkach, 8R012, Kwintet
Dostawca: FRISTADS KANSAS
1006505002XL 1006505003XL 100650500L 100650500M 100650500S 100650500XL 100650500XS 1006507302XL 1006507303XL 100650730L 100650730M 100650730S 100650730XL 100650730XS
KWIN1006505002XLEA 550 PLN
KWIN1006505002XL KWIN1006505003XL KWIN100650500L KWIN100650500M KWIN100650500S KWIN100650500XL KWIN100650500XS KWIN1006507302XL KWIN1006507303XL KWIN100650730L KWIN100650730M KWIN100650730S KWIN100650730XL KWIN100650730XS
Kombinezon do pomieszczeń czystych z mankietami na nogawkach, 8R012, Kwintet
Odzież robocza
These overalls are made of XR50 fabric consisting of 97% polyester and 3% conductive fibre. The fabric is ideal for sterile environments.
  • Fabric shows no change in pore size after 50 wash and autoclave cycles
  • Concealed zip, rounded neck with snap fastener
  • Elasticated cuffs, elasticated leg ends
  • Raglan sleeves
Weight: 115 g/m²