Drukuj
Łaźnie wodne, bez mieszania, seria JB Nova (Grant.)
Dostawca: GRANT INSTRUMENTS
JBN12 JBN18 JBN26 JBN5
462-8558EA 5290 PLN
462-8558 462-8559 462-1022 462-8557
Łaźnie wodne, bez mieszania, seria JB Nova (Grant.)
Łaźnie Łaźnie wodne bez mieszania
Digital unstirred water bath range with choice of models from 5 to 26 litres. All units have stainless steel tanks and durable painted steel cases, offer fast heat-up times, good temperature control, front panel lock, easy to use controls and a bright, clear display. All baths have heater mat technology ensuring optimum working space, without a heating element in the tank they are easier to clean. 12 litre and larger models feature a convenient drain tap on the front of the bath. The range is ideal for use in education as well as industrial applications such as product and materials testing.
  • Temperature stability ±0,5 °C, temperature range ambient 5 to 95 °C
  • Set and Forget™ technology. Modern technology for fast heat-up and excellent temperature accuracy
  • Baths have clear audible and visual alarms and advanced dry start and dry run protection, avoiding damage if accidentally switched on without water or bath runs dry. Simply fill and restart
  • User-defined single-point calibration allows optimum accuracy at a frequently used temperatures
  • LED display with 0,5 °C resolution
  • Front panel lock-out ensures current settings are not accidentally altered
*RT = Ambient
Dodatkowe informacje: Supplied with a transparent polycarbonate lid and perforated base tray which allows any container to be placed directly in the bath. UK and EU plug included.