Drukuj
Buty ochronne, sznurowane, Steelite™ Thor FW11, Portwest
Dostawca: Portwest
FW11BKR47 FW11BKR46 FW11BKR45 FW11BKR44 FW11BKR43 FW11BKR42 FW11BKR41 FW11BKR40 FW11BKR39 FW11BKR38
PRTWFW11BKR47EA 357 PLN
PRTWFW11BKR47 PRTWFW11BKR46 PRTWFW11BKR45 PRTWFW11BKR44 PRTWFW11BKR43 PRTWFW11BKR42 PRTWFW11BKR41 PRTWFW11BKR40 PRTWFW11BKR39 PRTWFW11BKR38
Buty ochronne, sznurowane, Steelite™ Thor FW11, Portwest
Obuwie i buty
Anti-static safety ankle boots made from full grain water resistant leather and a dual-density PU/TPU sole (petrol and chemical resistant).
  • Shock absorbent heel
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Certyfikaty: EN ISO 20345:2011; S3