Drukuj
Kolumny UHPLC, Pinnacle® DB IBD UHPLC (USP L68), Restek
Dostawca: Restek
9425252 9425212 9425232
554-1572EA 0 PLN
554-1572 554-1570 554-1571
Kolumny UHPLC, Pinnacle® DB IBD UHPLC (USP L68), Restek
Kolumny chromatograficzne
An intrinsically base-deactivated (IBD) phase, containing a polar group within, or intrinsic to, the hydrocarbon bonded phase. Unique selectivity and a high level of base deactivation, while reducing or eliminating the need for mobile phase additives.