Drukuj
Kolumny UHPLC, Pinnacle® DB C18 (USP L1), Restek
Dostawca: Restek
9414512-700 9414515-700 9414522-700 9414525-700 9414532-700 9414535-700 9414552-700 9414555-700 9414562-700 9414565-700 9414572-700 9414575-700 9414511 9414512 9414515 9414521 9414522 9414525 9414531 9414532 9414535 9414551 9414552 9414555 9414561 9414562 9414565 9414571 9414572 9414575 941450210 941450212 9414212_U 9414232 9414252_U 9414315 9414311 9414312 9414312-700 9414313-700 9414315-700 941431E 9414331 9414332 9414332-700 9414333-700 9414335 9414335-700 941433E 9414351 9414352 9414352-700 9414353-700 9414355 9414355-700 941435E 941451E 941452E 941453E 941455E 941456E 941457E 9414212 9414252
REST9414512-700EA 0 PLN
REST9414512-700 REST9414515-700 REST9414522-700 REST9414525-700 REST9414532-700 REST9414535-700 REST9414552-700 REST9414555-700 REST9414562-700 REST9414565-700 REST9414572-700 REST9414575-700 REST9414511 REST9414512 REST9414515 REST9414521 REST9414522 REST9414525 REST9414531 REST9414532 REST9414535 REST9414551 REST9414552 REST9414555 REST9414561 REST9414562 REST9414565 REST9414571 REST9414572 REST9414575 REST941450210 REST941450212 554-1558 554-1559 554-1560 REST9414315 REST9414311 REST9414312 REST9414312-700 REST9414313-700 REST9414315-700 REST941431E REST9414331 REST9414332 REST9414332-700 REST9414333-700 REST9414335 REST9414335-700 REST941433E REST9414351 REST9414352 REST9414352-700 REST9414353-700 REST9414355 REST9414355-700 REST941435E REST941451E REST941452E REST941453E REST941455E REST941456E REST941457E REST9414212 REST9414252
Kolumny UHPLC, Pinnacle® DB C18 (USP L1), Restek
Kolumny chromatograficzne
Highly base-deactivated spherical silica manufactured by Restek. Monomeric C18 bonding. Hydrophobic C18 phase suitable for analyses of a wide range of compounds, from acidic through slightly basic. Replaces Hypersil® BDS C18 and Pinnacle® ODS Amine.