Drukuj
Buty ochronne, sznurowane, Compositelite™ Trekker FC64, Portwest
Dostawca: Portwest
FC64BKR41 FC64BKR43 FC64BKR42 FC64BKR40 FC64BKR39 FC64BKR38 FC64BKR37 FC64BKR47 FC64BKR46 FC64BKR45 FC64BKR44
PRTWFC64BKR41EA 258 PLN
PRTWFC64BKR41 PRTWFC64BKR43 PRTWFC64BKR42 PRTWFC64BKR40 PRTWFC64BKR39 PRTWFC64BKR38 PRTWFC64BKR37 PRTWFC64BKR47 PRTWFC64BKR46 PRTWFC64BKR45 PRTWFC64BKR44
Buty ochronne, sznurowane, Compositelite™ Trekker FC64, Portwest
Obuwie i buty
Metal-free, anti-static, lightweight safety shoe made from cow suede leather upper plus a durable, dual-density, oil resistant sole.
  • Shock-absorbent heel
  • Composite toecap (200 J)
Colour: Grey
Certyfikaty: EN 20345:2011 + A1:2007; S1, SRC.