Drukuj
Półbuty ochronne, sznurowane, X-Light, 711033/711038, Abeba
Dostawca: Abeba
711033/35 711033/36 711033/37 711033/38 711033/39 711033/40 711033/41 711033/42 711033/43 711033/44 711033/45 711033/46 711033/47 711033/48 711038/35 711038/36 711038/37 711038/38 711038/39 711038/40 711038/41 711038/42 711038/43 711038/44 711038/45 711038/46 711038/47 711038/48
ABEB711033/35EA 402 PLN
ABEB711033/35 ABEB711033/36 ABEB711033/37 ABEB711033/38 ABEB711033/39 ABEB711033/40 ABEB711033/41 ABEB711033/42 ABEB711033/43 ABEB711033/44 ABEB711033/45 ABEB711033/46 ABEB711033/47 ABEB711033/48 ABEB711038/35 ABEB711038/36 ABEB711038/37 ABEB711038/38 ABEB711038/39 ABEB711038/40 ABEB711038/41 ABEB711038/42 ABEB711038/43 ABEB711038/44 ABEB711038/45 ABEB711038/46 ABEB711038/47 ABEB711038/48
Półbuty ochronne, sznurowane, X-Light, 711033/711038, Abeba
Obuwie i buty
Anti-static shoe made from smooth leather upper, reflective material, and a lightweight PU sole, resistant to fuels.
  • Perspiration-absorbent, Silver Point lining
  • Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbing system
  • Protective steel toecap (200 J)
  • Instep area with elastic band
Suitable for use in kitchens and in food processing industries.
Colour: White or black.
Certyfikaty: DGUV 112-191; EN ISO 20345:2011; S2, SRC.