Drukuj
System do badania parazytologicznego kału, ParasiTrap® AF, Biosepar
Dostawca: Biosepar
8486AF/50 8486AF/100 8484AF/50 8484AF/100 8484AF/250 8488AF/100 8490AF/50 8488AF/250 8488AF/50
8486AF/50EA 0 PLN
8486AF/50 8486AF/100 8484AF/50 8484AF/100 8484AF/250 8488AF/100 8490AF/50 8488AF/250 8488AF/50
System do badania parazytologicznego kału, ParasiTrap® AF, Biosepar
Systemy do diagnostyki klinicznej Clinical Reagents and Kits
ParasiTrap® enables detection of worm eggs, protozoa and larvae. The ParasiTrap® is a transport and testing system for the parasitological examination of stools in human and veterinary medicine. It is a completely closed system with 4 simple steps to make the diagnosis: 1. load tube with sample; 2. add solvent and stain; 3. filter and concentrate; 4. examine under microscope. It was developed to facilitate the unpleasant task of processing infectious stools in laboratory environments, reduce risk of infection and ensure the best possible yield of parasites. The closed system also allows macroscopic diagnostic procedures to be performed where necessary. This diagnostics concept fully complies with the modern quality management criteria of national and international guidelines (EU, CDC, WHO, BGA, DPG). All systems are free of merthiolate and ether. The ParasiTrap® AF-System contains a filled processing and transport tube I (with Medium AF) and the processing tube II with integrated stainless steel filter system. Accessories: separation medium B, staining solution medium C, Pasteur pipettes (disposable), spoons and cotton wool sticks. Additional screw caps for processing tube II.
  • High parasite yield and high diagnostic reliability
  • Outstanding quality of microscope viewing fields
  • Simple handling (examinations can be interrupted at any time, including for longer periods)
  • No cooling
  • Free of merthiolate (Thiomersal) and ether