Drukuj
System do parazytologicznego badania kału, ParasiTrap® Dual Eco, Biosepar
Dostawca: Biosepar
8486DUALECO 8486DUALECO/50 8486DUALECO/10
8486DUALECOEA 0 PLN
8486DUALECO 8486DUALECO/50 8486DUALECO/10
System do parazytologicznego badania kału, ParasiTrap® Dual Eco, Biosepar
Systemy do diagnostyki klinicznej Clinical Reagents and Kits
ParasiTrap Dual Eco contains filled processing and transport tubes I (50 pieces Medium A Bailenger and 50 pieces Medium A ECO) and 100 pieces processing tubes II with integrated stainless steel filter system.
Dodatkowe informacje: Separation medium B, staining solution medium C, Pasteur pipettes (disposable), spoons and cotton wool sticks. Additional screw caps for processing tube II.