Drukuj
System do badania parazytologicznego kału, ParasiTrap® Dual, Biosepar
Dostawca: Biosepar
8486DUAL 8486DUAL/50 8486DUAL/100
8486DUALEA 0 PLN
8486DUAL 8486DUAL/50 8486DUAL/100
System do badania parazytologicznego kału, ParasiTrap® Dual, Biosepar
Systemy do diagnostyki klinicznej Clinical Reagents and Kits
ParasiTrap Dual contains filled processing and transport tubes I (50 pieces Medium A Bailenger and 50 pieces Medium AF) and 100 pieces processing tubes II with integrated stainless steel filter system.
Dodatkowe informacje: Separation medium B, staining solution medium C, Pasteur pipettes (disposable), spoons and cotton wool sticks. Additional screw caps for processing tube II.