Drukuj
System do parazytologicznego badania kału, ParasiTrap® Eco, Biosepar
Dostawca: Biosepar
8484ECO/50 8484ECO/250 8490ECO/50 8488ECO/50 8488ECO/250 8488ECO/100 8486ECO/100 8486ECO/50 8488ECO/1000
8484ECO/50EA 931 PLN
8484ECO/50 8484ECO/250 8490ECO/50 8488ECO/50 8488ECO/250 8488ECO/100 8486ECO/100 8486ECO/50 8488ECO/1000
System do parazytologicznego badania kału, ParasiTrap® Eco, Biosepar
Systemy do diagnostyki klinicznej Clinical Reagents and Kits
ParasiTrap Eco contains a filled processing and transport tube I (with Medium A Eco) and the processing tube II with integrated stainless steel filter system.
  • High parasite yield and high diagnostic reliability
  • Outstanding quality of microscope viewing fields
  • Examinations can be interrupted at any time, including for longer periods
  • No cooling, high sensitivity, environmentally friendly
  • No formalin, no hazardous materials, free of ether
Dodatkowe informacje: Separation medium B, staining solution medium C, Pasteur pipettes (disposable), spoons and cotton wool sticks. Additional screw caps for processing tube II.