Drukuj
Kolumny HPLC, Hypersil GOLD™ AX, Thermo Fisher Scientific
Dostawca: Thermo Fisher Scientific
26103-012101 26103-102130
555-6511EA 2840 PLN
555-6511 555-6469
Kolumny HPLC, Hypersil GOLD™ AX, Thermo Fisher Scientific
Kolumny chromatograficzne
A novel polymeric ligand bonded to highly pure base deactivated silica.
  • Outstanding peak shape and selectivity
  • High stability under demanding pH conditions
  • Higher efficiency than polymer-based ion exchange columns
  • Improved speed with 1,9 µm particle size columns