Drukuj
Lejki do proszków, PP, SEMADENI
Dostawca: SEMADENI
0211 0213
SEMA0211EA 60.8 PLN
SEMA0211 SEMA0213
Lejki do proszków, PP, SEMADENI
Lejki, rozdzielacze i wkraplacze Lejki do materiałów sypkich
Funnels, made of PP, translucent. Used for transferring powders. Also suitable for liquids.
  • Parallel stem minimises powder bridging
  • Very good chemical and temperature resistance
  • Autoclavable