Drukuj
Anionowymienne kolumny chromatograficzne, HiTrap™ Q XL
Dostawca: Cytiva
17-5158-01 17-5159-01
17-5158-01EA 959 PLN
17-5158-01 17-5159-01
Anionowymienne kolumny chromatograficzne, HiTrap™ Q XL
Kolumny chromatograficzne
HiTrap Q XL are strong anion exchangers prepacked with Q Sepharose XL media, optimized for fast, convenient small-scale protein capture using ion exchange (IEX) chromatography.
  • High binding capacities with high flow rates
  • Convenient and affordable for fast, easy separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump, or chromatography system such as ÄKTA design
Q Sepharose XL and SP Sepharose XL resins have long chains of dextran coupled to a robust, 6% highly cross-linked agarose matrix. The dextran chains increase the exposure of the Q or SP charged groups, which results in very high loading capacities.