Drukuj

Perfluoropelargonic acid, Sigma-Aldrich®

Dostawca: Merck

Synonim: Perfluorononanoic acid

M-Clarity™ quality level = MQ100

394459-25G 394459-5G
SIAL394459-25GEA 704 PLN
SIAL394459-25G SIAL394459-5G
Perfluoropelargonic acid, Sigma-Aldrich®
Perfluoropelargonic acid
Formuła: C₉HF₁₇O₂
MW: 464.07 g/mol
Numer MDL: MFCD00039605
Numer CAS: 375-95-1
Numer WE (dawny EINECS): 206-801-3
Jednostka: 3261
ADR: 8 III

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR