Drukuj
Kolumienki SPE, CHROMABOND® PA
Dostawca: MACHEREY-NAGEL
730126 730127 730384 730007 731849 731851
MANA730126EA 1030 PLN
MANA730126 MANA730127 MANA730384 MANA730007 MANA731849 MANA731851
Kolumienki SPE, CHROMABOND® PA
Kolumny chromatograficzne
CHROMABOND® PA is a high purity polyamide 6 phase for SPE.
  • Medium polar phase
  • Separation mechanism mainly based on hydrogen bonds
Recommended for natural products like phenolic compounds, flavonoids and anthocyanins.