Drukuj
Spodnie malarskie, S817
Dostawca: Portwest
S817WHR1XL S817WHR2XL S817WHR3XL S817WHRL S817WHRM S817WHRS S817WHRXS
113-7573EA 198 PLN
113-7573 113-7574 113-7575 113-7576 113-7577 113-7578 113-7579
Spodnie malarskie, S817
Odzież robocza
Modern painters trousers made of 100% pre-shrunk cotton.
  • Knee pad pockets, hip and rule pockets
  • Multi-layered thigh pocket, mobile phone pocket
  • Elasticated back waist
Weight: 305 g/m²