Drukuj
Wkłady ekstrakcji do fazy stałej, HyperSep™ SAX
Dostawca: Thermo Fisher Scientific
60108-521 60108-434 60108-419 60108-360 60108-418 60108-417 60108-713 60108-714 60108-715
HYPE60108-521EA 1310 PLN
HYPE60108-521 HYPE60108-434 HYPE60108-419 HYPE60108-360 HYPE60108-418 HYPE60108-417 HYPE60108-713 HYPE60108-714 HYPE60108-715
Wkłady ekstrakcji do fazy stałej, HyperSep™ SAX
Kolumny chromatograficzne
Strong anion exchange sorbent for extraction of weak acids.
  • Extracts negatively charged compounds from both aqueous and non-aqueous solutions
  • Ideally suited for the extraction of weak acids, e.g. carboxylic acids
  • Amine group masks the effect of the carbon chain of the functional group
  • Selectivity can be tuned by selection of buffer