Drukuj
Kombinezony odporne na chemikalia, 4540+
Dostawca: 3M
4540PLUSM 4540PLUSL 4540PLUSXL 4540PLUSXXL 4540PLUSS 4540PLUS3XL
113-6304EA 101 PLN
113-6304 113-6303 113-6306 113-6307 113-6305 113-6301
Kombinezony odporne na chemikalia, 4540+
Odzież jednorazowa
These disposable overalls are made of a high quality laminated material and provide a barrier protection against light liquid splashes and hazardous dusts.
  • Elasticated waist and ankles for freedom of movement and knitted cuffs for extra comfort
  • Breathable back panel (SMMMS) for reduced heat stress and increased comfort
  • Three-panel hood design for a better fit and compatibility with other PPE
  • Two-way zip and sealable storm flap for enhanced protection
  • Low-linting properties and anti-static coating
Typical applications may include paint spraying, industrial and pharmaceutical processing, asbestos inspection, insulation laying, and vehicle and machine maintenance (oils and greases).
Colour: White/blue
Certyfikaty: Type 5/6, EN 1149-5, EN 1073-2:2002; Cat. III.